Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenemethanol, α-[(butylamino)methyl]-4-hydroxy-, sulfate (2:1) | 5716-20-1 | 99.5% | 25 MG
SDP

Catalog No. 5000413904
Encompass_Preferred

Bamethan hemisulfate is a vasodilator that enhances blood flow and reduces vascular resistance. It improves oxygen delivery to tissues and aids in the management of hypertension through its relaxing effects on blood vessels.

  • Molecular weight: 307.36
  • Formula: C12H21NO6S
  • Appearance: Solid
  • Color: White to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Store at room temperature, keep dry and cool. In solvent, store at -80°C for 1 year or -20°C for 6 months.

Catalog No. 50-004-13904 Supplier Medchemexpress LLC Supplier No. 25MGHYB1538A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bamethan hemisulfate is a vasodilator that enhances blood flow and reduces vascular resistance. It improves oxygen delivery to tissues and aids in the management of hypertension through its relaxing effects on blood vessels.

  • Molecular weight: 307.36
  • Formula: C12H21NO6S
  • Appearance: Solid
  • Color: White to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Store at room temperature, keep dry and cool. In solvent, store at -80°C for 1 year or -20°C for 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.