Learn More
Medchemexpress LLC Benzenepropanamide, a-[[(1,2-dihydro-2-oxo-6-quinolinyl)sulfonyl)amino]-N-(2-furanylmethyl)-N-(2-thienylmethyl)- | 1681056-62-1 | 98.0% | 547.65 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY148113
Encompass_Preferred
PG 34 is a weak O-GlcNAc transferase (OGT) inhibitor with an IC50 of 68 μM and a close structural analog of OSMI-1. In vitro studies show that PG34, at concentrations up to 50 μM for 24 hours, does not detectably inhibit OGT in CHO cells, nor does it affect apoptotic signaling. A moderate effect on cell viability was observed only at the highest concentration of 50 μM.
  • Weak O-GlcNAc transferase (OGT) inhibitor
  • IC50 of 68 μM
  • Structural analog of OSMI-1
  • Does not inhibit OGT in CHO cells (0.5-50 μM)
  • No effect on apoptotic signaling
  • Moderate effect on cell viability at 50 μM
  • No reduction in global O-GlcNAcylation (10, 25, 50 μM)
Catalog No. 50-003-62670