Learn More
Medchemexpress LLC Benzenepropanamide, α-[(cyclohexylcarbonyl)amino]-N-[5-[[(1,1-dimethylethyl)amino]sulfonyl]-1-naphth | 2241025-68-1 | 99.8% | 535.70 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112438100MG
Encompass_Preferred
MF-094 is a potent and selective USP30 inhibitor with an IC50 of 120 nM. It increases protein ubiquitination and accelerates mitophagy. It is for research use only and not sold to patients.
  • Potent and selective USP30 inhibitor
  • IC50 of 120 nM for USP30
  • Increases protein ubiquitination
  • Accelerates mitophagy
  • <30% inhibitory activity for a panel of 22 USPs assays at 10 μM
Catalog No. 50-004-38620