Learn More
Medchemexpress LLC Benzenepropanamide, α-[(cyclohexylcarbonyl)amino]-N-[5-[[(1,1-dimethylethyl)amino]sulfonyl]-1-naphthalenyl]-, (aS)- | 2241025-68-1 | 99.8% | 535.70 | 25 MG
SDP
Catalog No. 5000439838
Encompass_Preferred
MF-094 is a potent and selective USP30 inhibitor with an IC50 of 120 nM. It functions by increasing protein ubiquitination and accelerating mitophagy. This product is intended for research use only and is not sold to patients.
  • Potent and selective USP30 inhibitor with an IC50 of 120 nM.
  • Increases protein ubiquitination and accelerates mitophagy.
  • Shows less than 30% inhibitory activity for a panel of 22 USPs assays at 10 μM.
  • Purity of 99.75% with an enantiomeric excess (ee) of 100.00%.
  • Offered in various quantities and forms.
  • Includes data sheet, COA, SDS, and handling instructions.
Catalog No. 50-004-39838 Supplier Medchemexpress LLC Supplier No. 25MGHY112438
MF-094 is a potent and selective USP30 inhibitor with an IC50 of 120 nM. It functions by increasing protein ubiquitination and accelerating mitophagy. This product is intended for research use only and is not sold to patients.
  • Potent and selective USP30 inhibitor with an IC50 of 120 nM.
  • Increases protein ubiquitination and accelerates mitophagy.
  • Shows less than 30% inhibitory activity for a panel of 22 USPs assays at 10 μM.
  • Purity of 99.75% with an enantiomeric excess (ee) of 100.00%.
  • Offered in various quantities and forms.
  • Includes data sheet, COA, SDS, and handling instructions.