Learn More
Medchemexpress LLC Benzenepropanamine, 2-chloro-N-[(1R)-1-(3-methoxyphenyl)ethyl] hydrochloride | 177172-49-5 | MFCD16038895 | 99.7% | 340.29 g/mol | C18H23Cl2NO | 25 MG
SDP
Catalog No. 5000248475
Encompass_Preferred
Tecalcet hydrochloride (R-568 hydrochloride) is an orally active calcimimetic that allosterically and positively modulates the calcium-sensing receptor (CaSR), increasing sensitivity to extracellular calcium. Supplied as the hydrochloride salt for research use, it is characterized by defined purity and analytical data.
  • Orally active calcimimetic for CaSR-related studies.
  • Positive allosteric modulator of the calcium-sensing receptor (CaSR).
  • High purity as confirmed by analytical testing.
  • Supplied in small research pack sizes suitable for dosing and formulation.
  • Molecular formula C18H23Cl2NO; molecular weight 340.29 g/mol.
Catalog No. 50-002-48475 Supplier Medchemexpress LLC Supplier No. HY10167A25MG
Tecalcet hydrochloride (R-568 hydrochloride) is an orally active calcimimetic that allosterically and positively modulates the calcium-sensing receptor (CaSR), increasing sensitivity to extracellular calcium. Supplied as the hydrochloride salt for research use, it is characterized by defined purity and analytical data.
  • Orally active calcimimetic for CaSR-related studies.
  • Positive allosteric modulator of the calcium-sensing receptor (CaSR).
  • High purity as confirmed by analytical testing.
  • Supplied in small research pack sizes suitable for dosing and formulation.
  • Molecular formula C18H23Cl2NO; molecular weight 340.29 g/mol.