Learn More
Medchemexpress LLC Benzenepropanamine, γ-(2-methoxyphenoxy)-N-methyl- | 53179-07-0 | 98.56% | 50 MG
SDP
Catalog No. 5000411441
Encompass_Preferred
Nisoxetine is a potent and selective inhibitor of noradrenaline transporter (NET), with a Kd of 0.76 nM. It is an antidepressant and local anesthetic, capable of blocking voltage-gated sodium channels. It is suitable for laboratory research and experimentation in various biological activities.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Kd of 0.76 nM (NET)
  • Antidepressant properties
  • Local anesthetic properties
  • Blocks voltage-gated sodium channels
Catalog No. 50-004-11441 Supplier Medchemexpress LLC Supplier No. HYB170450MG
Nisoxetine is a potent and selective inhibitor of noradrenaline transporter (NET), with a Kd of 0.76 nM. It is an antidepressant and local anesthetic, capable of blocking voltage-gated sodium channels. It is suitable for laboratory research and experimentation in various biological activities.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Kd of 0.76 nM (NET)
  • Antidepressant properties
  • Local anesthetic properties
  • Blocks voltage-gated sodium channels