Learn More
Medchemexpress LLC Benzenepropanamine, γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride (1:1) | 57226-68-3 | 99.58% | 331.76 | 10 MG
SDP
Catalog No. 5000452104
Encompass_Preferred
Norfluoxetine hydrochloride is an active N-demethylated metabolite of Fluoxetine. Fluoxetine is a selective serotonin (5-HT) reuptake inhibitor that is metabolized to Norfluoxetine hydrochloride by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4. Norfluoxetine hydrochloride inhibits 5-HT uptake and inhibits CaV3.3 T current (IC50 = 5 μM). It also possesses anticonvulsant activity.
  • Inhibits 5-HT uptake.
  • Inhibits CaV3.3 T current (IC50 = 5 μM).
  • Exhibits anticonvulsant activity.
Catalog No. 50-004-52104 Supplier Medchemexpress LLC Supplier No. HYW01123510MG
Norfluoxetine hydrochloride is an active N-demethylated metabolite of Fluoxetine. Fluoxetine is a selective serotonin (5-HT) reuptake inhibitor that is metabolized to Norfluoxetine hydrochloride by cytochrome P450 (CYP) 2D6, CYP2C19, and CYP3A4. Norfluoxetine hydrochloride inhibits 5-HT uptake and inhibits CaV3.3 T current (IC50 = 5 μM). It also possesses anticonvulsant activity.
  • Inhibits 5-HT uptake.
  • Inhibits CaV3.3 T current (IC50 = 5 μM).
  • Exhibits anticonvulsant activity.