Learn More
Medchemexpress LLC Benzenepropanamine, gamma-(2-methoxyphenoxy)-N-methyl-, hydrochloride | 57754-86-6 | 99.9% | 307.82 | C17H22ClNO2 | 10 MG
SDP
Catalog No. 5000297802
Encompass_Preferred
Nisoxetine hydrochloride is a research-grade small molecule that potently and selectively inhibits the noradrenaline (norepinephrine) transporter (NET) and can block voltage-gated sodium channels. It is supplied as a solid powder for use in transporter pharmacology, neurotransmitter uptake assays, and sodium channel studies.
  • Potent, selective noradrenaline transporter (NET) inhibitor (reported Kd ≈ 0.76 nM).
  • Acts as an antidepressant-class compound and local anesthetic via sodium channel blockade.
  • High purity suitable for biochemical assays and pharmacology studies.
  • Available in small-scale powder sizes and DMSO solution formats for ready use.
  • Stable when stored under recommended conditions (powder and in solvent).
Catalog No. 50-002-97802 Supplier Medchemexpress LLC Supplier No. HYB1704A10MG
Nisoxetine hydrochloride is a research-grade small molecule that potently and selectively inhibits the noradrenaline (norepinephrine) transporter (NET) and can block voltage-gated sodium channels. It is supplied as a solid powder for use in transporter pharmacology, neurotransmitter uptake assays, and sodium channel studies.
  • Potent, selective noradrenaline transporter (NET) inhibitor (reported Kd ≈ 0.76 nM).
  • Acts as an antidepressant-class compound and local anesthetic via sodium channel blockade.
  • High purity suitable for biochemical assays and pharmacology studies.
  • Available in small-scale powder sizes and DMSO solution formats for ready use.
  • Stable when stored under recommended conditions (powder and in solvent).