Learn More
Medchemexpress LLC Benzenepropanoic acid, a-oxo- | 156-06-9 | 99.9% | 164.16 | 1 ML
SDP
Catalog No. 5000453777
Encompass_Preferred
Phenylpyruvic acid is a precursor of the antifungal compound phenyllactic acid. It can improve the antifungal activity of eight lactic acid bacterial strains when added to a defined growth medium. Phenylpyruvic acid demonstrates improved inhibitory activity against fungal bread contaminants such as Aspergillus niger and Penicillium roqueforti. It affects enzyme activity of the pentose phosphate pathway involved in the oxidative phase in rat brain homogenates and can reduce glucose-6-phosphate dehydrogenase activity.
  • Precursor of antifungal compound phenyllactic acid.
  • Improves antifungal activity of lactic acid bacterial strains.
  • Demonstrates improved inhibitory activity against fungal bread contaminants (Aspergillus niger and Penicillium roqueforti).
  • Affects enzyme activity of the pentose phosphate pathway in rat brain homogenates.
  • Reduces glucose-6-phosphate dehydrogenase activity.
Catalog No. 50-004-53777 Supplier Medchemexpress LLC Supplier No. HYW01253010MM/1ML
Phenylpyruvic acid is a precursor of the antifungal compound phenyllactic acid. It can improve the antifungal activity of eight lactic acid bacterial strains when added to a defined growth medium. Phenylpyruvic acid demonstrates improved inhibitory activity against fungal bread contaminants such as Aspergillus niger and Penicillium roqueforti. It affects enzyme activity of the pentose phosphate pathway involved in the oxidative phase in rat brain homogenates and can reduce glucose-6-phosphate dehydrogenase activity.
  • Precursor of antifungal compound phenyllactic acid.
  • Improves antifungal activity of lactic acid bacterial strains.
  • Demonstrates improved inhibitory activity against fungal bread contaminants (Aspergillus niger and Penicillium roqueforti).
  • Affects enzyme activity of the pentose phosphate pathway in rat brain homogenates.
  • Reduces glucose-6-phosphate dehydrogenase activity.