Learn More
Medchemexpress LLC Benzenepropanoic acid, ß-(aminomethyl)-4-chloro-, hydrochloride (1:1) | 28311-31-1 | 99.8% | C10H13Cl2NO2 | 1 ML
SDP
Catalog No. 5000432533
Encompass_Preferred
Baclofen hydrochloride is a lipophilic derivative of γ-aminobutyric acid (GABA) and an orally active, selective metabotropic GABAB receptor (GABABR) agonist. It mimics the action of GABA, producing slow presynaptic inhibition through the GABAB receptor. This compound exhibits high blood-brain barrier penetrance, making it suitable for muscle spasticity research.
  • Orally active, selective GABAB receptor agonist.
  • Mimics GABA action for slow presynaptic inhibition.
  • Exhibits high blood-brain barrier penetrance.
  • Increases cell viability in huntingtin-expressing striatal cells.
  • Enhances chymotrypsin-like proteasome activity.
  • Ameliorates motor deficits in Huntington's disease animal models.
  • Reduces neuronal intranuclear inclusions in animal models.
Catalog No. 50-004-32533 Supplier Medchemexpress LLC Supplier No. HYB0007C10MM/1ML
Baclofen hydrochloride is a lipophilic derivative of γ-aminobutyric acid (GABA) and an orally active, selective metabotropic GABAB receptor (GABABR) agonist. It mimics the action of GABA, producing slow presynaptic inhibition through the GABAB receptor. This compound exhibits high blood-brain barrier penetrance, making it suitable for muscle spasticity research.
  • Orally active, selective GABAB receptor agonist.
  • Mimics GABA action for slow presynaptic inhibition.
  • Exhibits high blood-brain barrier penetrance.
  • Increases cell viability in huntingtin-expressing striatal cells.
  • Enhances chymotrypsin-like proteasome activity.
  • Ameliorates motor deficits in Huntington's disease animal models.
  • Reduces neuronal intranuclear inclusions in animal models.