Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzeneruthenium(II) Chloride Dimer

Catalog No. 3415685G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-156-85G 5 g
34-156-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 34-156-85G Supplier Sigma Aldrich Supplier No. 3415685G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrosilylation CatalystsCatalyst for SNAr reactions of fluoroarenes and cyclic amines (eq. 1)Precatalyst for the hydrogenation of +a-keto esters (eq. 2)Precursor for the preparation of other ruthenium catalysts (eq. 3)

Specifications

CAS 37366-09-9
Molecular Formula C12H12Cl4Ru2
Linear Formula Ru2(C6H6)2Cl4
Quantity 5 g
MDL Number MFCD00064686
Synonym (Benzene)dichlororuthenium dimer; Benzeneruthenium dichloride dimer; Bis(benzene)tetrachlorodiruthenium; Bis[(benzene)dichlororuthenium]; Bis[(benzene)dichlororuthenium]; Dibenzenetetrachlorodiruthenium
Molecular Weight (g/mol) 500.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.