Learn More
Medchemexpress LLC Benzenesulfinic acid, sodium salt (1:1) | 873-55-2 | 99.97% | 164.16 | 250 G
SDP
Catalog No. 5000454039
Encompass_Preferred
Sodium benzenesulfinate is a reagent used in organic synthesis to create other compounds, such as sulfones. It undergoes rhodium-catalyzed desulfinative coupling with aldehydes to yield ketones.
  • Reagent for organic synthesis
  • Used to create sulfones
  • Facilitates rhodium-catalyzed desulfinative coupling with aldehydes
  • Yields ketones
Catalog No. 50-004-54039 Supplier Medchemexpress LLC Supplier No. 250GHYW013598
Sodium benzenesulfinate is a reagent used in organic synthesis to create other compounds, such as sulfones. It undergoes rhodium-catalyzed desulfinative coupling with aldehydes to yield ketones.
  • Reagent for organic synthesis
  • Used to create sulfones
  • Facilitates rhodium-catalyzed desulfinative coupling with aldehydes
  • Yields ketones