Learn More
Apexbio Technology LLC Benzenesulfonamide 98-10-2 100mg
SDP
Catalog No. 502240664
Encompass_Preferred
Benzenesulfonamide (CAS 98-10-2) is an amide derivative of benzenesulfonic acid widely utilized as a synthetic intermediate for pharmaceuticals and other chemicals Mechanistically benzenesulfonamide derivatives have been investigated for their inhibitory activity against various human carbonic anhydrases (CAs) with certain 4-substituted diazo derivatives exhibiting strong binding affinity particularly for CA I and II isoforms as demonstrated by isothermal titration calorimetry and fluorescence-based thermal shift assays In in vivo models indole-based benzenesulfonamide derivatives show oral activity in rat and sheep models of airway inflammation attenuating physiological responses to antigenic or cholinergic challenge Benzenesulfonamide remains in preclinical evaluation
This item is not returnable. View return policy
Catalog No. 50-224-0664 Supplier Apexbio Technology LLC Supplier No. B6154100
Benzenesulfonamide (CAS 98-10-2) is an amide derivative of benzenesulfonic acid widely utilized as a synthetic intermediate for pharmaceuticals and other chemicals Mechanistically benzenesulfonamide derivatives have been investigated for their inhibitory activity against various human carbonic anhydrases (CAs) with certain 4-substituted diazo derivatives exhibiting strong binding affinity particularly for CA I and II isoforms as demonstrated by isothermal titration calorimetry and fluorescence-based thermal shift assays In in vivo models indole-based benzenesulfonamide derivatives show oral activity in rat and sheep models of airway inflammation attenuating physiological responses to antigenic or cholinergic challenge Benzenesulfonamide remains in preclinical evaluation