Learn More
Medchemexpress LLC Benzenesulfonamide, 4-[5-phenyl-3-(3-(4-(trifluoromethyl)phenyl)carbamoyl)propyl]-1H-pyrazol-1-yl | 1287761-01-6 | 98.1% | 50 MG
SDP
Catalog No. 5000233662
Encompass_Preferred
PTUPB is a research-use small molecule that potently and selectively inhibits soluble epoxide hydrolase (sEH) and cyclooxygenase-2 (COX-2). It is used in preclinical pharmacology to study inflammation and tumor biology.
  • Dual sEH and COX-2 inhibition (sEH IC50 = 0.9 nM; COX-2 IC50 = 1.26 μM).
  • High chemical purity (98.05% by LCMS).
  • Molecular weight 543.56 g/mol; formula C26H24F3N5O3S.
  • Supplied as a powder in a 50 MG pack size.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-33662 Supplier Medchemexpress LLC Supplier No. HY12259150MG
PTUPB is a research-use small molecule that potently and selectively inhibits soluble epoxide hydrolase (sEH) and cyclooxygenase-2 (COX-2). It is used in preclinical pharmacology to study inflammation and tumor biology.
  • Dual sEH and COX-2 inhibition (sEH IC50 = 0.9 nM; COX-2 IC50 = 1.26 μM).
  • High chemical purity (98.05% by LCMS).
  • Molecular weight 543.56 g/mol; formula C26H24F3N5O3S.
  • Supplied as a powder in a 50 MG pack size.
  • For research use only; not for human or clinical use.