Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenesulfonamide, 4-[(6-amino-4-pyrimidinyl)amino]-, hydrochloride (1:1) | 21886-12-4 | 99.6% | 301.75 | 25 MG
SDP

Catalog No. 5000440052
Encompass_Preferred

PNU112455A hydrochloride is an ATP-competitive inhibitor of CDK2 and CDK5. It binds to the ATP site of CDK2 and CDK5 with Kms of 3.6 and 3.2 μM, respectively. This product is for research use only and is not sold to patients.

  • ATP-competitive CDK2 and CDK5 inhibitor.
  • Binds to the ATP site of CDK2 and CDK5.
  • Purity: 99.56%.
  • Available in various sizes and concentrations, including solid and solution forms.
  • Soluble in DMSO and H2O.
  • Provided with detailed solubility and storage information for both in vitro and in vivo applications.

Catalog No. 50-004-40052 Supplier Medchemexpress LLC Supplier No. 25MGHY112468
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PNU112455A hydrochloride is an ATP-competitive inhibitor of CDK2 and CDK5. It binds to the ATP site of CDK2 and CDK5 with Kms of 3.6 and 3.2 μM, respectively. This product is for research use only and is not sold to patients.

  • ATP-competitive CDK2 and CDK5 inhibitor.
  • Binds to the ATP site of CDK2 and CDK5.
  • Purity: 99.56%.
  • Available in various sizes and concentrations, including solid and solution forms.
  • Soluble in DMSO and H2O.
  • Provided with detailed solubility and storage information for both in vitro and in vivo applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.