Learn More
Medchemexpress LLC Benzenesulfonamide, 4-amino-N-2-thiazolyl- | 72-14-0 | 99.7% | 500 MG
SDP
Catalog No. 5000408275
Encompass_Preferred
Sulfathiazole is an orally active, organosulfur compound functioning as a short-acting sulfonamide antibiotic and an endocrine disruptor. It targets the steroidogenic pathway, enhancing CYP19 activity in human adrenal cells (H295R) and upregulating mRNA expression of CYP17, CYP19, and 3β-HSD. It also increases 17-estradiol (E2) production and exhibits endocrine-disrupting effects on aquatic organisms like the Japanese medaka fish. This product is a white to off-white solid with a molecular formula of C9H9N3O2S2 and a molecular weight of 255.32. It is intended for research use only.
  • Purity of 99.71%
  • Orally active sulfonamide antibiotic
  • Targets steroidogenic pathway by enhancing CYP19 activity
  • Appears as a white to off-white solid
  • Recommended for long-term storage in powder form at -20°C for 3 years or 4°C for 2 years
  • Recommended for storage in solution at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-08275 Supplier Medchemexpress LLC Supplier No. HYB0507500MG
Sulfathiazole is an orally active, organosulfur compound functioning as a short-acting sulfonamide antibiotic and an endocrine disruptor. It targets the steroidogenic pathway, enhancing CYP19 activity in human adrenal cells (H295R) and upregulating mRNA expression of CYP17, CYP19, and 3β-HSD. It also increases 17-estradiol (E2) production and exhibits endocrine-disrupting effects on aquatic organisms like the Japanese medaka fish. This product is a white to off-white solid with a molecular formula of C9H9N3O2S2 and a molecular weight of 255.32. It is intended for research use only.
  • Purity of 99.71%
  • Orally active sulfonamide antibiotic
  • Targets steroidogenic pathway by enhancing CYP19 activity
  • Appears as a white to off-white solid
  • Recommended for long-term storage in powder form at -20°C for 3 years or 4°C for 2 years
  • Recommended for storage in solution at -80°C for 6 months or -20°C for 1 month