Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenesulfonamide, 4-amino-N-(5-chloro-2-quinoxalinyl) | 97919-22-7 | 99.1% | 334.78 g/mol | C14H11ClN4O2S | 100 MG
SDP

Catalog No. 5000242017
Encompass_Preferred

Chloroquinoxaline sulfonamide (CAS 97919-22-7) is a research reagent that acts as a topoisomerase II α/β poison with reported antiparasitic and antitumor activity. It is supplied as a solid for laboratory use and is soluble in DMSO.

  • Functions as a topoisomerase II alpha/beta poison.
  • Exhibits antiparasitic and antitumor activity in research studies.
  • Molecular formula C14H11ClN4O2S; molecular weight 334.78 g/mol.
  • Soluble in DMSO: 125 mg/mL (approx. 373.38 mM).
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Typical purity provided: 99.06% by HPLC.

Catalog No. 50-002-42017 Supplier Medchemexpress LLC Supplier No. HY106662100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloroquinoxaline sulfonamide (CAS 97919-22-7) is a research reagent that acts as a topoisomerase II α/β poison with reported antiparasitic and antitumor activity. It is supplied as a solid for laboratory use and is soluble in DMSO.

  • Functions as a topoisomerase II alpha/beta poison.
  • Exhibits antiparasitic and antitumor activity in research studies.
  • Molecular formula C14H11ClN4O2S; molecular weight 334.78 g/mol.
  • Soluble in DMSO: 125 mg/mL (approx. 373.38 mM).
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Typical purity provided: 99.06% by HPLC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.