Learn More
Apexbio Technology LLC Benzenesulfonamide 98-10-2 500mg
SDP
Catalog No. 5000088382
Encompass_Preferred
Benzenesulfonamide (CAS 98-10-2) is an amide derivative of benzenesulfonic acid widely utilized as a synthetic intermediate for pharmaceuticals and other chemicals Mechanistically benzenesulfonamide derivatives have been investigated for their inhibitory activity against various human carbonic anhydrases (CAs) with certain 4-substituted diazo derivatives exhibiting strong binding affinity particularly for CA I and II isoforms as demonstrated by isothermal titration calorimetry and fluorescence-based thermal shift assays In in vivo models indole-based benzenesulfonamide derivatives show oral activity in rat and sheep models of airway inflammation attenuating physiological responses to antigenic or cholinergic challenge Benzenesulfonamide remains in preclinical evaluation
Catalog No. 50-000-88382 Supplier Apexbio Technology LLC Supplier No. B6154500
Benzenesulfonamide (CAS 98-10-2) is an amide derivative of benzenesulfonic acid widely utilized as a synthetic intermediate for pharmaceuticals and other chemicals Mechanistically benzenesulfonamide derivatives have been investigated for their inhibitory activity against various human carbonic anhydrases (CAs) with certain 4-substituted diazo derivatives exhibiting strong binding affinity particularly for CA I and II isoforms as demonstrated by isothermal titration calorimetry and fluorescence-based thermal shift assays In in vivo models indole-based benzenesulfonamide derivatives show oral activity in rat and sheep models of airway inflammation attenuating physiological responses to antigenic or cholinergic challenge Benzenesulfonamide remains in preclinical evaluation