Learn More
Medchemexpress LLC Benzenesulfonate sodium | 515-42-4 | MFCD00065179 | 99.4% | 180.16 g/mol | C6H5NaO3S | 1000 G
SDP
Catalog No. 5000301145
Encompass_Preferred
Benzenesulfonate sodium is the sodium salt of benzenesulfonic acid supplied as a white to off-white solid reagent for laboratory and analytical use. It has CAS 515-42-4, molecular formula C6H5NaO3S, and a molecular weight of 180.16 g/mol. Typical purity is approximately 99.4% and bulk pack sizes are available.
  • White to off-white solid suitable for reagent use.
  • High purity supports analytical and reference applications.
  • Water-soluble source of the benzenesulfonate anion.
  • Useful as a buffer component or analytical standard.
  • Available in bulk sizes for large-scale laboratory use.
Catalog No. 50-003-01145 Supplier Medchemexpress LLC Supplier No. HYW0165101000G
Benzenesulfonate sodium is the sodium salt of benzenesulfonic acid supplied as a white to off-white solid reagent for laboratory and analytical use. It has CAS 515-42-4, molecular formula C6H5NaO3S, and a molecular weight of 180.16 g/mol. Typical purity is approximately 99.4% and bulk pack sizes are available.
  • White to off-white solid suitable for reagent use.
  • High purity supports analytical and reference applications.
  • Water-soluble source of the benzenesulfonate anion.
  • Useful as a buffer component or analytical standard.
  • Available in bulk sizes for large-scale laboratory use.