Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzenesulfonic Acid

Catalog No. 126351KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-635-1KGF 1 kg
12-635-25GF 25 g
12-635-100GF 100 g
12-635-500GF 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 12-635-1KGF Supplier Sigma Aldrich Supplier No. 126351KGF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzenesulfonic acid is an aryl sulfonic acid that can be used to form an organic salt with 5,7-dimethyl-1,8-naphthyridine-2-amine, that can be a synthon for developing supramolecular structures. It can also act as a dopant for the polymerization of pyrrole to form poly(pyrrole), a conducting polymer useful in the development of flexible capacitors.

Specifications

CAS 98-11-3
Molecular Formula C6H6O3S
Linear Formula C6H5SO3H
MDL Number MFCD00011689
Quantity 1 kg
Synonym Phenylsulfonic acid
Molecular Weight (g/mol) 158.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.