Learn More
Medchemexpress LLC Benzenesulfonic acid, 2,2'-[(9,10-dihydro-9,10-dioxo-1,4-anthracenediyl)diimino]bis[5-methyl-, sodium salt (1:2) | 4403-90-1 | 82.8% | 622.58 | 100 MG
SDP
Catalog No. 5000428177
Encompass_Preferred
Alizarin cyanin green F is an alizarin dye that reacts with calcium possibly via sulfonic acid and/or OH groups.
  • Functions as an alizarin dye.
  • Reacts with calcium, potentially through sulfonic acid and/or OH groups.
Catalog No. 50-004-28177 Supplier Medchemexpress LLC Supplier No. HYD1175100MG
Alizarin cyanin green F is an alizarin dye that reacts with calcium possibly via sulfonic acid and/or OH groups.
  • Functions as an alizarin dye.
  • Reacts with calcium, potentially through sulfonic acid and/or OH groups.