Learn More
Medchemexpress LLC Benzenesulfonic acid, 4-[2-[4-(dimethylamino)phenyl]diazenyl]-, sodium salt (1:1) | 547-58-0 | MFCD00007502 | 99.9% | 327.34 g/mol | C14H14N3NaO3S | 10 G
SDP
Catalog No. 5000228588
Encompass_Preferred
Methyl Orange is a soluble azo dye used as an acid-base indicator and staining reagent for laboratory titrations and pH testing. Supplied as an analytical-grade sodium salt, it is provided in solid packages for preparation of standard solutions.
  • Analytical-grade purity suitable for laboratory use.
  • Soluble sodium salt for straightforward dissolution in aqueous media.
  • Distinct color change in the acidic pH range useful for titrations and assays.
  • Available in small solid packages for solution preparation.
  • Molecular weight 327.34 g/mol for accurate solution calculations.
Catalog No. 50-002-28588 Supplier Medchemexpress LLC Supplier No. HY11890710G
Methyl Orange is a soluble azo dye used as an acid-base indicator and staining reagent for laboratory titrations and pH testing. Supplied as an analytical-grade sodium salt, it is provided in solid packages for preparation of standard solutions.
  • Analytical-grade purity suitable for laboratory use.
  • Soluble sodium salt for straightforward dissolution in aqueous media.
  • Distinct color change in the acidic pH range useful for titrations and assays.
  • Available in small solid packages for solution preparation.
  • Molecular weight 327.34 g/mol for accurate solution calculations.