Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonyl Chloride, Spectrum™ Chemical
SDP

Catalog No. 18620303 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-303 1 L
18-620-302 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-303 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B20031LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-09-9

Specifications

Boiling Point 252°C/485.6°F
Quantity 1 L
Molecular Weight (g/mol) 176.62
Percent Purity 98.5%
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Benzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.