Learn More
Medchemexpress LLC Benzenesulphonamide | 98-10-2 | MFCD00007930 | 99.5% | 157.19 g/mol | C6H7NO2S | 10 G
SDP
Catalog No. 5000213487
Encompass_Preferred
Benzenesulphonamide is an organic sulfonamide used as a biochemical reagent and analytical standard. It has formula C6H7NO2S, molecular weight 157.19 g/mol, and CAS number 98-10-2. The compound is supplied as a white to off-white solid with reported purity of about 99.5% and is employed in enzyme inhibition studies and analytical assays.
Catalog No. 50-002-13487 Supplier Medchemexpress LLC Supplier No. HY10108710G
Benzenesulphonamide is an organic sulfonamide used as a biochemical reagent and analytical standard. It has formula C6H7NO2S, molecular weight 157.19 g/mol, and CAS number 98-10-2. The compound is supplied as a white to off-white solid with reported purity of about 99.5% and is employed in enzyme inhibition studies and analytical assays.