Learn More
Medchemexpress LLC Benzenesulphonamide | 98-10-2 | MFCD00007930 | 99.5% | 157.19 g/mol | C6H7NO2S | 100 G
SDP
Catalog No. 5000213343
Encompass_Preferred
Benzenesulphonamide is an analytical standard and research reagent (CAS 98-10-2) used in biochemical assays, including carbonic anhydrase II inhibition studies. It has molecular formula C6H7NO2S, molecular weight 157.19 g/mol, and reported purity around 99.5%. Available in solid and DMSO solution formats; consult the product datasheet and safety data sheet for storage, handling, and application details.
  • High reported purity suitable for analytical and research applications.
  • Available as solid and as DMSO solutions for flexible workflows.
  • Supplied with a product data sheet and safety data sheet for compliance and safe handling.
  • Applicable for carbonic anhydrase II inhibition studies and related assays.
  • Well characterized: formula C6H7NO2S, molecular weight 157.19 g/mol.
Catalog No. 50-002-13343 Supplier Medchemexpress LLC Supplier No. HY101087100G
Benzenesulphonamide is an analytical standard and research reagent (CAS 98-10-2) used in biochemical assays, including carbonic anhydrase II inhibition studies. It has molecular formula C6H7NO2S, molecular weight 157.19 g/mol, and reported purity around 99.5%. Available in solid and DMSO solution formats; consult the product datasheet and safety data sheet for storage, handling, and application details.
  • High reported purity suitable for analytical and research applications.
  • Available as solid and as DMSO solutions for flexible workflows.
  • Supplied with a product data sheet and safety data sheet for compliance and safe handling.
  • Applicable for carbonic anhydrase II inhibition studies and related assays.
  • Well characterized: formula C6H7NO2S, molecular weight 157.19 g/mol.