Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzenesulphonamide | 98-10-2 | MFCD00007930 | 99.5% | 157.19 g/mol | C6H7NO2S | 5 G
SDP

Catalog No. 5000213287
Encompass_Preferred

Benzenesulphonamide is a high-purity analytical standard for biochemical and analytical research, commonly used as a carbonic anhydrase inhibitor reference and as a reagent for assay development and method validation. The compound is supplied as a solid in multiple pack sizes and is accompanied by safety and technical documentation.

  • High purity for analytical applications (~99.5%).
  • CAS number 98-10-2 for unambiguous identification.
  • Molecular weight 157.19 g/mol.
  • Chemical formula C6H7NO2S.
  • Supplied as a solid, available in small pack sizes including 5 G.
  • Suitable as a reference standard for enzyme inhibition and assay work.
  • Provided with a safety data sheet for handling and storage.

Catalog No. 50-002-13287 Supplier Medchemexpress LLC Supplier No. HY1010875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzenesulphonamide is a high-purity analytical standard for biochemical and analytical research, commonly used as a carbonic anhydrase inhibitor reference and as a reagent for assay development and method validation. The compound is supplied as a solid in multiple pack sizes and is accompanied by safety and technical documentation.

  • High purity for analytical applications (~99.5%).
  • CAS number 98-10-2 for unambiguous identification.
  • Molecular weight 157.19 g/mol.
  • Chemical formula C6H7NO2S.
  • Supplied as a solid, available in small pack sizes including 5 G.
  • Suitable as a reference standard for enzyme inhibition and assay work.
  • Provided with a safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.