Learn More
Medchemexpress LLC Benzenesulphonamide | 98-10-2 | MFCD00007930 | 99.5% | 157.19 g/mol | C6H7NO2S | 50 G
SDP
Catalog No. 5000213481
Encompass_Preferred
Benzenesulphonamide is a small-molecule carbonic anhydrase inhibitor used in preclinical research on Alzheimer's disease, epilepsy, inflammation, and several cancer types.
  • Exhibits carbonic anhydrase II inhibitory activity.
  • High purity (~99.5%).
  • Available as a solid and in DMSO solution formats.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • White to off-white solid appearance.
  • Analytical documentation available, including COA, SDS, NMR, and LC-MS.
  • Stable at room temperature for years; in solvent store frozen for long-term stability.
Catalog No. 50-002-13481 Supplier Medchemexpress LLC Supplier No. HY10108750G
Benzenesulphonamide is a small-molecule carbonic anhydrase inhibitor used in preclinical research on Alzheimer's disease, epilepsy, inflammation, and several cancer types.
  • Exhibits carbonic anhydrase II inhibitory activity.
  • High purity (~99.5%).
  • Available as a solid and in DMSO solution formats.
  • Soluble in DMSO at 100 mg/mL; may require sonication.
  • White to off-white solid appearance.
  • Analytical documentation available, including COA, SDS, NMR, and LC-MS.
  • Stable at room temperature for years; in solvent store frozen for long-term stability.