Learn More
Selleck Chemical LLC Benzethonium Chloride S4162-50mg
SDP
Catalog No. 503109094
Encompass_Preferred
Benzethonium chloride is a potent inhibitor of nAChRs it inhibits 4 2 nAChRs and 7 nAChRs with IC50 of 49 nM and 122 nM respectively
Catalog No. 50-310-9094 Supplier Selleck Chemical LLC Supplier No. S416250MG
Benzethonium chloride is a potent inhibitor of nAChRs it inhibits 4 2 nAChRs and 7 nAChRs with IC50 of 49 nM and 122 nM respectively