Learn More
Medchemexpress LLC Benzethonium chloride (Standard) | 121-54-0 | 98.3% | 50 MG
SDP
Catalog No. 5000408830
Encompass_Preferred
Benzethonium chloride (Standard) is the analytical standard of Benzethonium chloride, intended for research and analytical applications. It inhibits nicotinic acetylcholine receptors in human recombinant α7 and α4β2 neurons in Xenopus laevis oocytes. This product is for research use only and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Inhibits nicotinic acetylcholine receptors in human recombinant α7 and α4β2 neurons
  • Suitable for use in HPLC, GC, and MS experiments
  • For research use only
Catalog No. 50-004-08830 Supplier Medchemexpress LLC Supplier No. HYB0942R50MG
Benzethonium chloride (Standard) is the analytical standard of Benzethonium chloride, intended for research and analytical applications. It inhibits nicotinic acetylcholine receptors in human recombinant α7 and α4β2 neurons in Xenopus laevis oocytes. This product is for research use only and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Inhibits nicotinic acetylcholine receptors in human recombinant α7 and α4β2 neurons
  • Suitable for use in HPLC, GC, and MS experiments
  • For research use only