Learn More
Medchemexpress LLC Benzethonium-d7 chloride | 121-54-0 | 98.6% | 1 MG
SDP
Catalog No. 5000408020
Encompass_Preferred
Benzethonium-d7 chloride is a deuterium-labeled form of Benzethonium chloride. It inhibits human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors in Xenopus oocytes. This compound is suitable for various analytical applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium (2H, D) stable isotope-labeled inhibitor
  • Suitable for in vivo cell experiments
  • Used in therapeutic drug monitoring
Catalog No. 50-004-08020 Supplier Medchemexpress LLC Supplier No. HYB0942S1MG
Benzethonium-d7 chloride is a deuterium-labeled form of Benzethonium chloride. It inhibits human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors in Xenopus oocytes. This compound is suitable for various analytical applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium (2H, D) stable isotope-labeled inhibitor
  • Suitable for in vivo cell experiments
  • Used in therapeutic drug monitoring