Learn More
Medchemexpress LLC Benzethonium-d7 chloride | 121-54-0 | 98.6% | 455.12 g/mol | C27H35D7ClNO2 | 10 MG
SDP
Catalog No. 5000297729
Encompass_Preferred
Benzethonium-d7 chloride is the deuterium-labeled analogue of benzethonium chloride supplied as a solid for analytical and research use. It serves as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS, and is used in studies of neuronal nicotinic acetylcholine receptor activity. The compound has a molecular weight of 455.12 g/mol and high listed purity (98.64%).
  • Deuterium-labeled internal standard for analytical methods
  • Suitable for NMR, GC-MS, and LC-MS quantitation
  • High purity (98.6%) for reliable results
  • Solid form for easy handling and storage
  • Molecular weight 455.12 g/mol
  • Available in small, research-scale pack sizes
Catalog No. 50-002-97729 Supplier Medchemexpress LLC Supplier No. HYB0942S10MG
Benzethonium-d7 chloride is the deuterium-labeled analogue of benzethonium chloride supplied as a solid for analytical and research use. It serves as a tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS, and is used in studies of neuronal nicotinic acetylcholine receptor activity. The compound has a molecular weight of 455.12 g/mol and high listed purity (98.64%).
  • Deuterium-labeled internal standard for analytical methods
  • Suitable for NMR, GC-MS, and LC-MS quantitation
  • High purity (98.6%) for reliable results
  • Solid form for easy handling and storage
  • Molecular weight 455.12 g/mol
  • Available in small, research-scale pack sizes