Learn More
Medchemexpress LLC Benzethonium-d7 chloride | 121-54-0 | 98.6% | 5 MG
SDP
Catalog No. 5000408952
Encompass_Preferred
Benzethonium-d7 chloride is a deuterium-labeled version of Benzethonium chloride. It is known to inhibit human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors in Xenopus oocytes. This compound is valuable as a tracer and internal standard for quantitative analysis in drug development processes.
  • Deuterium-labeled compound for tracer applications
  • Inhibits human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Incorporates stable heavy isotopes for quantitation during drug development
Catalog No. 50-004-08952 Supplier Medchemexpress LLC Supplier No. HYB0942S5MG
Benzethonium-d7 chloride is a deuterium-labeled version of Benzethonium chloride. It is known to inhibit human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors in Xenopus oocytes. This compound is valuable as a tracer and internal standard for quantitative analysis in drug development processes.
  • Deuterium-labeled compound for tracer applications
  • Inhibits human recombinant α7 and α4β2 neuronal nicotinic acetylcholine receptors
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Incorporates stable heavy isotopes for quantitation during drug development