Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzetimide hydrochloride | 5633-14-7 | 99.4% | 1 ML
SDP

Catalog No. 5000411941
Encompass_Preferred

Benzetimide hydrochloride is a muscarinic acetylcholine receptor (mAChR) antagonist, also identified as R4929 and Dioxatrine. This product is intended for research use only and is not for human use.

  • Acts as a muscarinic acetylcholine receptor (mAChR) antagonist.
  • Suitable for various research applications including neurological, eye or ear, neurodegenerative diseases, and Parkinson's disease.
  • Useful in GPCR/G Protein, neuronal signaling, and mAChR studies.
  • High purity of 99.44%.
  • Molecular weight of 398.93.
  • Appearance: solid, white to off-white.
  • Soluble in DMSO and water.

Catalog No. 50-004-11941 Supplier Medchemexpress LLC Supplier No. HYB1547A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzetimide hydrochloride is a muscarinic acetylcholine receptor (mAChR) antagonist, also identified as R4929 and Dioxatrine. This product is intended for research use only and is not for human use.

  • Acts as a muscarinic acetylcholine receptor (mAChR) antagonist.
  • Suitable for various research applications including neurological, eye or ear, neurodegenerative diseases, and Parkinson's disease.
  • Useful in GPCR/G Protein, neuronal signaling, and mAChR studies.
  • High purity of 99.44%.
  • Molecular weight of 398.93.
  • Appearance: solid, white to off-white.
  • Soluble in DMSO and water.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.