Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzetimide hydrochloride | 5633-14-7 | MFCD00210783 | 99.4% | 398.93 | 10 MG
SDP

Catalog No. 5000365513
Encompass_Preferred

Benzetimide hydrochloride is an antagonist of the muscarinic acetylcholine receptor (mAChR). It is intended for research use only and is not sold to patients.

  • Target: mAChR
  • Appearance: Solid
  • Color: White to off-white
  • Related classifications include neurological, eye or ear disease, neurodegenerative disease, Parkinson's disease, GPCR/G protein, and neuronal signaling.

Catalog No. 50-003-65513 Supplier Medchemexpress LLC Supplier No. 10MGHYB1547A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzetimide hydrochloride is an antagonist of the muscarinic acetylcholine receptor (mAChR). It is intended for research use only and is not sold to patients.

  • Target: mAChR
  • Appearance: Solid
  • Color: White to off-white
  • Related classifications include neurological, eye or ear disease, neurodegenerative disease, Parkinson's disease, GPCR/G protein, and neuronal signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.