Learn More
Cayman Chemical Benzetimide (hydrochloride), Purity: ≥95%, CAS: 5633-14-7, 100 mg
SDP
Catalog No. 502950456
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist; induces mydriasis and inhibits pilocarpine-induced salivation and lacrimation in rats (ED50s = 0.06 and 0.04 mg/kg, respectively)
Catalog No. 50-295-0456 Supplier Cayman Chemical Supplier No. 34527100 MG
A muscarinic acetylcholine receptor antagonist; induces mydriasis and inhibits pilocarpine-induced salivation and lacrimation in rats (ED50s = 0.06 and 0.04 mg/kg, respectively)