Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzhydrylamine

Catalog No. A5360525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5360525G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5360525G Supplier Sigma Aldrich Supplier No. A5360525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromopyridin-3-yl sulfurofluoridate can be used in the pd-catalyzed suzuki-miyuara cross-coupling reaction
  • The fluorosulfate functionality was shown by sharpless and coworkers to have distinct reactivity in comparison to other functional groups used in suzuki-miyuara reactions, such as bromides, chlorides and triflates
  • This inherent property provides a strong potential for selective suzuki-miyuara couplings for chemoselective reaction sequences.palladium-catalyzed, ligand-free suzuki reaction in water using aryl fluorosulfates

Specifications

CAS 91-00-9
Melting Point 12°C (lit.)
Density 1.063 g/mL (at 25°C (literature))
Boiling Point 295°C (lit.)
Molecular Formula C13H13N
Refractive Index n20/D 1.595 (literature)
Linear Formula (C6H5)2 CHNH2
MDL Number MFCD00008059
Quantity 500 mg
Synonym alpha-Aminodiphenylmethane
Molecular Weight (g/mol) 183.25
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.