Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzidine, MilliporeSigma™ Supelco™

Catalog No. 111019972
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9972 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-9972 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 31614100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 92-87-5
Physical Form Neat
Linear Formula C12H12N2
MDL Number MFCD00039765
Synonym 4,4 ′-Diaminobiphenyl
Type Analytical Standard
Quantity 100 mg
Formula Weight 184.24
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.