Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzil, 98%

Catalog No. aaa1257922
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1257922 100 g
AAA1257936 500 g
AAA125790E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1257922 Supplier Thermo Scientific Chemicals Supplier No. A1257922
Only null left
Add to Cart

CAS: 134-81-6 | C14H10O2 | 210.23 g/mol

Benzil is used as a pharmaceutical intermediate and uv curing resin photosensitizer. In polymer chemistry, it is used as a photoinitiator. Further, it serves as a potent inhibitor of human carboxylesterases. It is used in the preparation of diketimines by reacting with amines. It is also involved in the famous benil-benzilic acid rearrangement. In addition this, it reacts with 1,3-diphenylacetone to get tetraphenylcyclopentadienone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzil is used as a pharmaceutical intermediate and uv curing resin photosensitizer. In polymer chemistry, it is used as a photoinitiator. Further, it serves as a potent inhibitor of human carboxylesterases. It is used in the preparation of diketimines by reacting with amines. It is also involved in the famous benil-benzilic acid rearrangement. In addition this, it reacts with 1,3-diphenylacetone to get tetraphenylcyclopentadienone.

Solubility
Soluble in ethanol and ethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-81-6
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.23
MDL Number MFCD00003080
InChI Key WURBFLDFSFBTLW-UHFFFAOYSA-N
Synonym benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
PubChem CID 8651
ChEBI CHEBI:51507
IUPAC Name 1,2-diphenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 94°C to 96°C
Density 1.521
Boiling Point 346°C to 348°C
Quantity 100 g
Beilstein 608047
Merck Index 14,1078
Solubility Information Soluble in ethanol and ethyl ether. Insoluble in water.
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material Benzil
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-157-0
RTECSNumber DD1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.