Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzimidazole, 98%

Catalog No. AC186420025
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186420025 2.5 kg Plastic drum
AC186420250 25 g Plastic bottle
AC186421000 100 g Plastic bottle
AC186425000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC186420025 Supplier Thermo Scientific Chemicals Supplier No. 186420025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51-17-2 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
IUPAC Name 1H-benzimidazole
SMILES N1C=NC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 174.0°C
Color Beige to Brown
Boiling Point >360.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Quantity 2.5 kg
Beilstein 23, 131
Merck Index 15, 1083
Formula Weight 118.14
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Benzimidazole, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-081-4
RTECSNumber DD5425000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.