Learn More
Benzimidazole, 99%
Description
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 170°C to 174°C |
| Boiling Point | 360°C |
| Quantity | 1000 g |
| Beilstein | 109682 |
| Merck Index | 14,1081 |
| Solubility Information | Sparingly soluble in water. |
| IUPAC Name | 1H-benzimidazole |
| Formula Weight | 118.14 |
| Percent Purity | 99% |
| Chemical Name or Material | Benzimidazole |
Safety and Handling
| EINECSNumber | 200-081-4 |
| RTECSNumber | DD5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.