Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzimidazole, 99%

Catalog No. AAA1276318
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1276318 50 g
AAA1276330 250 g
AAA127630B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1276318 Supplier Thermo Scientific Chemicals Supplier No. A1276318
Add to Cart

CAS: 51-17-2 | C7H6N2 | 118.14 g/mol

Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole is a building block for making anthelmintic drugs like albendazole, mebendazole and triclabendazole and anti-ulcer drugs like pantoprazole, rabeprazole, lansoprazole and omeprazole. It is used in the preparation of pyrene derived benzimidazole linked polymers (BILPs), which are used for selective carbon dioxide separation applications due to their properties like porous materials, regenerability and sorbent selection parameters.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-17-2
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005585
InChI Key HYZJCKYKOHLVJF-UHFFFAOYSA-N
Synonym benzimidazole, 1h-benzo d imidazole, 1,3-benzodiazole, benzoimidazole, o-benzimidazole, 3-azaindole, benziminazole, benzoglyoxaline, azindole, 1,3-diazaindene
PubChem CID 5798
ChEBI CHEBI:41275
SMILES N1C=NC2=CC=CC=C12

Specifications

Melting Point 170°C to 174°C
Boiling Point 360°C
Quantity 50 g
Beilstein 109682
Merck Index 14,1081
Solubility Information Sparingly soluble in water.
IUPAC Name 1H-1,3-benzodiazole
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material Benzimidazole
EINECSNumber 200-081-4
RTECSNumber DD5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.