Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzine (Petroleum Naphtha), Fisher Chemical

Catalog No. B26420 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Steel Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B264-20 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B264-20

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Benzine (Petroleum Naphtha)
CAS 64742-89-8, 1330-20-7, 108-38-3, 100-41-4, 71-43-2
Color Colorless
Density 0.740g/cm³
Boiling Point 118.5°C
Flash Point 14°C
Packaging Steel Pail
Molecular Formula C5 to C10Hydrocarbons
Quantity 20 L
MDL Number MFCD00081849
UN Number 1268
Synonym Naphtha
Vapor Density 4.1g/mol
Vapor Pressure 15mmHg at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.