Learn More
Cayman Chemical Benzisoxazole Hsp90 Inhibitor, Purity: ≥98%, CAS: 1012788-65-6, 5 mg
SDP
Catalog No. 502887744
Encompass_Preferred
An Hsp90 inhibitor (IC50 = 30 nM in a fluorescence polarization assay); selective for Hsp90 over a panel of kinases (IC50s = >20 µM); inhibits the proliferation of HCT116, SK-BR-3, LNCaP, DU145, and H157 cancer cells (IC50s = 0.28, 0.37, 0.17, 0.29, and 0.23 µM, respectively); induces degradation of HER2 and the androgen receptor in SK-BR-3 and LNCaP cells, respectively (IC50 = 0.33 µM for both)
Catalog No. 50-288-7744 Supplier Cayman Chemical Supplier No. 100126945 MG
An Hsp90 inhibitor (IC50 = 30 nM in a fluorescence polarization assay); selective for Hsp90 over a panel of kinases (IC50s = >20 µM); inhibits the proliferation of HCT116, SK-BR-3, LNCaP, DU145, and H157 cancer cells (IC50s = 0.28, 0.37, 0.17, 0.29, and 0.23 µM, respectively); induces degradation of HER2 and the androgen receptor in SK-BR-3 and LNCaP cells, respectively (IC50 = 0.33 µM for both)