Learn More
Cayman Chemical BenznIdazole 100mg
SDP
Catalog No. 502940428
Encompass_Preferred
An orally bioavailable antiprotozoal agent; inhibits the growth of the parasites T. cruzi, T. vaginalis, G. lamblia, and E. histolytica (IC50s = 8.1, 18.62, 22.58, and 4.27 μM, respectively); inhibits clonogenic growth of human C33A cervical and KNS42 glioblastoma cancer cells under hypoxic, but not normoxic, conditions at 100 μM; decreases T. cruzi blood parasitemia to below detectable levels in a mouse model of chronic stable Chagas disease at 100 mg/kg per day
Catalog No. 50-294-0428 Supplier Cayman Chemical Supplier No. 29090100 MG
An orally bioavailable antiprotozoal agent; inhibits the growth of the parasites T. cruzi, T. vaginalis, G. lamblia, and E. histolytica (IC50s = 8.1, 18.62, 22.58, and 4.27 μM, respectively); inhibits clonogenic growth of human C33A cervical and KNS42 glioblastoma cancer cells under hypoxic, but not normoxic, conditions at 100 μM; decreases T. cruzi blood parasitemia to below detectable levels in a mouse model of chronic stable Chagas disease at 100 mg/kg per day