Learn More
Medchemexpress LLC Benznidazole | 22994-85-0 | 99.9% | 50 MG
SDP
Catalog No. 5000411456
Encompass_Preferred
Benznidazole (Ro 07-1051) is an antiparasitic medication, with an IC50 of 20.35 μM for Colombian T. cruzi strains, and has been used in the treatment of Chagas disease. It is for research use only and not sold to patients.
  • Antiparasitic medication
  • Used in the treatment of Chagas disease
  • IC50 of 20.35 μM for Colombian T. cruzi strains
  • For research use only
Catalog No. 50-004-11456 Supplier Medchemexpress LLC Supplier No. HYB154850MG
Benznidazole (Ro 07-1051) is an antiparasitic medication, with an IC50 of 20.35 μM for Colombian T. cruzi strains, and has been used in the treatment of Chagas disease. It is for research use only and not sold to patients.
  • Antiparasitic medication
  • Used in the treatment of Chagas disease
  • IC50 of 20.35 μM for Colombian T. cruzi strains
  • For research use only