Learn More
Medchemexpress LLC Benzo-15-crown-5-ether | 14098-44-3 | 99.0% | 268.31 | 5 G
SDP
Catalog No. 5000451600
Encompass_Preferred
Benzo-15-crown-5-ether can be used as a chelating agent for sodium and potassium. It is for research use only and not sold to patients.
  • Acts as a chelating agent for sodium and potassium.
  • For research use only.
Catalog No. 50-004-51600 Supplier Medchemexpress LLC Supplier No. HYW0085855G
Benzo-15-crown-5-ether can be used as a chelating agent for sodium and potassium. It is for research use only and not sold to patients.
  • Acts as a chelating agent for sodium and potassium.
  • For research use only.