Learn More
Medchemexpress LLC Benzo[a]phenazin-5-ol | 2089-82-9 | 99.5% | 25 MG
SDP
Catalog No. 5000230652
Encompass_Preferred
sAJM589 is a small-molecule c-Myc inhibitor that disrupts the Myc-Max heterodimer and is supplied as a powdered research chemical (C16H10N2O, MW 246.26 g/mol) with high reported purity and a reported IC50 of 1.8 μM for Myc. Store powders frozen or refrigerated per supplier recommendations.
  • Disrupts Myc-Max heterodimer, inhibiting c-Myc activity.
  • Reported IC50 of 1.8 μM for Myc.
  • High purity (99.47%), suitable for in vitro research.
  • Available as a powder in multiple solid sizes and as a DMSO solution format.
  • Stable when stored frozen according to supplier guidelines.
Catalog No. 50-002-30652 Supplier Medchemexpress LLC Supplier No. HY12268325MG
sAJM589 is a small-molecule c-Myc inhibitor that disrupts the Myc-Max heterodimer and is supplied as a powdered research chemical (C16H10N2O, MW 246.26 g/mol) with high reported purity and a reported IC50 of 1.8 μM for Myc. Store powders frozen or refrigerated per supplier recommendations.
  • Disrupts Myc-Max heterodimer, inhibiting c-Myc activity.
  • Reported IC50 of 1.8 μM for Myc.
  • High purity (99.47%), suitable for in vitro research.
  • Available as a powder in multiple solid sizes and as a DMSO solution format.
  • Stable when stored frozen according to supplier guidelines.