Learn More
Medchemexpress LLC Benzo[a]pyrene | 50-32-8 | MFCD00003602 | 100.0% | 252.31 g/mol | C20H12 | 50 MG
SDP
Catalog No. 5000240863
Encompass_Preferred
Benzo[a]pyrene is a polycyclic aromatic hydrocarbon used as a research reagent to model carcinogenesis and to study DNA damage and repair mechanisms. It forms DNA adducts and demonstrates lung carcinogenicity in animal models; handle with appropriate laboratory safety measures.
  • High purity suitable for analytical and biological studies.
  • Used to model DNA adduct formation and carcinogenesis in in vitro and in vivo assays.
  • Available as solid and solution formats for flexible dosing.
  • Molecular formula C20H12 and molecular weight 252.31 g/mol.
  • Safety data sheet and technical documentation available.
  • For research use only; not for diagnostic or therapeutic use.
Catalog No. 50-002-40863 Supplier Medchemexpress LLC Supplier No. HY10737750MG
Benzo[a]pyrene is a polycyclic aromatic hydrocarbon used as a research reagent to model carcinogenesis and to study DNA damage and repair mechanisms. It forms DNA adducts and demonstrates lung carcinogenicity in animal models; handle with appropriate laboratory safety measures.
  • High purity suitable for analytical and biological studies.
  • Used to model DNA adduct formation and carcinogenesis in in vitro and in vivo assays.
  • Available as solid and solution formats for flexible dosing.
  • Molecular formula C20H12 and molecular weight 252.31 g/mol.
  • Safety data sheet and technical documentation available.
  • For research use only; not for diagnostic or therapeutic use.