Learn More
Medchemexpress LLC Benzo[a]pyrene | 50-32-8 | MFCD00003602 | ≥98.0% | 252.31 | C20H12 | 1 G
SDP
Catalog No. 5000240861
Encompass_Preferred
Benzo[a]pyrene (CAS 50-32-8) is a polycyclic aromatic hydrocarbon used in laboratory research to model carcinogenesis and to evaluate chemopreventive agents. It is commonly employed as an analytical standard and reagent for studies of lung carcinogenicity in animal models.
  • High purity (≥98.0%) suitable for analytical and research use.
  • Molecular formula C20H12 and molecular weight 252.31 for accurate stoichiometry.
  • Intended for use in carcinogenesis and chemoprevention studies.
  • Available in multiple small pack sizes for laboratory experiments.
  • Includes a safety data sheet with handling and hazard information.
Catalog No. 50-002-40861 Supplier Medchemexpress LLC Supplier No. HY1073771G
Benzo[a]pyrene (CAS 50-32-8) is a polycyclic aromatic hydrocarbon used in laboratory research to model carcinogenesis and to evaluate chemopreventive agents. It is commonly employed as an analytical standard and reagent for studies of lung carcinogenicity in animal models.
  • High purity (≥98.0%) suitable for analytical and research use.
  • Molecular formula C20H12 and molecular weight 252.31 for accurate stoichiometry.
  • Intended for use in carcinogenesis and chemoprevention studies.
  • Available in multiple small pack sizes for laboratory experiments.
  • Includes a safety data sheet with handling and hazard information.