Learn More
Medchemexpress LLC Benzo[a]pyrene | 50-32-8 | MFCD00003602 | 99.8% | 252.31 g/mol | C20H12 | 5 MG
SDP
Catalog No. 5000241387
Encompass_Preferred
Benzo[a]pyrene is an analytical reference standard of the polycyclic aromatic hydrocarbon class, intended for research and analytical applications such as method development, calibration, and chemoprevention studies. The material is supplied as a light yellow to green-yellow solid in small pack sizes appropriate for laboratory use.
  • Analytical reference standard for calibration and method development.
  • High reported purity (99.8%) for reliable quantitation.
  • Chemical formula C20H12 and molecular weight 252.31 g/mol.
  • Available in a 5 mg pack size suitable for small-scale laboratory work.
  • Light yellow to green-yellow solid appearance for easy identification.
  • Intended for research and analytical applications including chemoprevention studies.
  • Store under conditions specified in the certificate of analysis.
Catalog No. 50-002-41387 Supplier Medchemexpress LLC Supplier No. HY107377R5MG
Benzo[a]pyrene is an analytical reference standard of the polycyclic aromatic hydrocarbon class, intended for research and analytical applications such as method development, calibration, and chemoprevention studies. The material is supplied as a light yellow to green-yellow solid in small pack sizes appropriate for laboratory use.
  • Analytical reference standard for calibration and method development.
  • High reported purity (99.8%) for reliable quantitation.
  • Chemical formula C20H12 and molecular weight 252.31 g/mol.
  • Available in a 5 mg pack size suitable for small-scale laboratory work.
  • Light yellow to green-yellow solid appearance for easy identification.
  • Intended for research and analytical applications including chemoprevention studies.
  • Store under conditions specified in the certificate of analysis.